Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 16 - 17 | 2250 | 90 | ||
NM_070018.6 | NP_502419.2 | 11 - 12 | 1518 | 90 | ||
GenBank mRNA | AB055112.1 | - | 13 - 14 | 1915 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |