Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 21 - 22 | 2910 | 125 | ||
NM_070018.6 | NP_502419.2 | 16 - 17 | 2178 | 125 | ||
GenBank mRNA | AB055112.1 | - | 18 - 19 | 2575 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |