Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 11 - 12 | 1655 | 118 | ||
NM_070018.6 | NP_502419.2 | 6 - 7 | 923 | 118 | ||
GenBank mRNA | AB055112.1 | - | 8 - 9 | 1320 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |