Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268734.3 | NP_001255663.1 | 14 - 15 | 2850 | 60 | |
NM_001268735.4 | NP_001255664.1 | 13 - 14 | 2829 | 60 | ||
NM_001268736.3 | NP_001255665.1 | 12 - 13 | 2793 | 60 | ||
NM_001268737.4 | NP_001255666.1 | 14 - 15 | 2884 | 60 | ||
NM_001268738.3 | NP_001255667.1 | 13 - 14 | 2853 | 60 | ||
NM_001268739.3 | NP_001255668.1 | 5 - 6 | 1485 | 60 | ||
NM_001268740.3 | NP_001255669.1 | 5 - 6 | 1485 | 60 | ||
NM_001306973.3 | NP_001293902.1 | 15 - 16 | 2886 | 60 | ||
NM_001306975.3 | NP_001293904.1 | 14 - 15 | 2865 | 60 | ||
NM_001306977.3 | NP_001293906.1 | 13 - 14 | 2829 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |