Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028037.4 | NP_001023208.1 | 10 - 11 | 2385 | 97 | |
NM_001306987.4 | NP_001293916.1 | 11 - 12 | 2334 | 97 | ||
GenBank mRNA | AY090019.1 | - | 10 - 11 | 2408 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |