Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028036.4 | NP_001023207.1 | 5 - 6 | 782 | 62 | |
NM_001028037.4 | NP_001023208.1 | 5 - 6 | 776 | 62 | ||
NM_001306987.4 | NP_001293916.1 | 6 - 7 | 725 | 62 | ||
NM_001380497.1 | NP_001368118.1 | 6 - 7 | 725 | 62 | ||
GenBank mRNA | AY090019.1 | - | 5 - 6 | 799 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |