Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scl-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070107.2 | NP_502508.1 | 1 - 2 | 62 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |