Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372945.2 | NP_001360500.1 | 3 - 4 | 406 | 76 | |
NM_001372946.3 | NP_001360501.1 | 3 - 4 | 406 | 76 | ||
NM_070143.4 | NP_502544.1 | 3 - 4 | 409 | 76 | ||
NM_070144.6 | NP_502545.1 | 3 - 4 | 398 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |