Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csn-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268796.4 | NP_001255725.1 | 1 - 2 | 261 | 68 | |
NM_001268797.2 | NP_001255726.1 | 1 - 2 | 257 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |