Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csn-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268796.4 | NP_001255725.1 | 4 - 5 | 1136 | 78 | |
NM_001268797.2 | NP_001255726.1 | 4 - 5 | 1117 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |