Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssu-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268875.3 | NP_001255804.1 | 4 - 5 | 666 | 109 | |
NM_001268877.3 | NP_001255806.1 | 4 - 5 | 749 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |