Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268956.1 | NP_001255885.1 | 1 - 2 | 348 | 107 | |
NR_066102.1 | 1 - 2 | 348 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |