Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268980.3 | NP_001255909.1 | 6 - 7 | 825 | 66 | |
NM_001268982.3 | NP_001255911.1 | 1 - 2 | 79 | 66 | ||
NM_001372971.2 | NP_001360155.1 | 6 - 7 | 825 | 66 | ||
GenBank mRNA | EU051652.1 | - | 6 - 7 | 841 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |