Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380283.2 | NP_001368373.1 | 14 - 15 | 2191 | 72 | |
NM_068617.8 | NP_501018.1 | 19 - 20 | 2970 | 72 | ||
GenBank mRNA | AF480439.1 | - | 19 - 20 | 2965 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |