Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hil-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026112.4 | NP_001021283.1 | 4 - 5 | 1446 | 65 | |
GenBank mRNA | AF216291.1 | - | 1 - 2 | 115 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |