Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068639.7 | NP_501040.1 | 3 - 4 | 324 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |