Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cogc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068691.7 | NP_501092.1 | 2 - 3 | 335 | 87 | |
GenBank mRNA | Z34976.1 | - | 333 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |