Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070773.2 | NP_503174.1 | 5 - 6 | 882 | 98 | |
NR_132474.1 | 4 - 5 | 1190 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |