Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070814.5 | NP_503215.2 | 5 - 6 | 906 | 68 | |
GenBank mRNA | AY204171.1 | - | 5 - 6 | 915 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |