Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070827.1 | NP_503228.1 | 3 - 4 | 521 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |