Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313121.3 | NP_001300050.1 | 1 - 2 | 214 | 176 | |
NM_070915.4 | NP_503316.1 | 1 - 2 | 214 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |