Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ergo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313235.3 | NP_001300164.1 | 1 - 2 | 821 | 68 | |
NM_070961.9 | NP_503362.2 | 4 - 5 | 2238 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |