Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srab-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313374.1 | NP_001300303.1 | 4 - 5 | 586 | 75 | |
NM_071093.2 | NP_503494.1 | 5 - 6 | 852 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |