Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071195.2 | NP_503596.2 | 4 - 5 | 630 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |