Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029097.5 | NP_001024268.1 | 2 - 3 | 166 | 75 | |
NM_001029098.5 | NP_001024269.1 | 1 - 2 | 103 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |