Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038398.3 | NP_001033487.1 | 5 - 6 | 982 | 82 | |
NM_001316801.3 | NP_001303730.1 | 3 - 4 | 521 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |