Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071223.8 | NP_503624.2 | 1 - 2 | 257 | 71 | |
GenBank mRNA | AF273807.1 | - | 1 - 2 | 224 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |