Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129385.4 | NP_001122857.1 | 2 - 3 | 165 | 74 | |
NM_071334.2 | NP_503735.1 | 2 - 3 | 152 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |