Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316818.1 | NP_001303747.1 | 1 - 2 | 192 | 135 | |
NM_001316819.1 | NP_001303748.1 | 1 - 2 | 192 | 135 | ||
NM_001373005.1 | NP_001360560.1 | 1 - 2 | 192 | 135 | ||
NM_071355.2 | NP_503756.2 | 1 - 2 | 192 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |