Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071366.1 | NP_503767.1 | 4 - 5 | 828 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |