Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071633.1 | NP_504034.1 | 3 - 4 | 920 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |