Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-34A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047606.5 | NP_001041071.1 | 2 - 3 | 517 | 107 | |
NM_001047607.4 | NP_001041072.1 | 2 - 3 | 476 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |