Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028998.4 | NP_001024169.1 | 1 - 2 | 130 | 69 | |
NM_001380634.1 | NP_001367924.1 | 1 - 2 | 130 | 69 | ||
NM_001380635.1 | NP_001367923.1 | 1 - 2 | 132 | 69 | ||
NM_001383298.1 | 1 - 2 | 124 | 69 | |||
GenBank mRNA | AF427474.1 | - | 1 - 2 | 152 | 69 | |
AF427475.1 | - | 1 - 2 | 152 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |