Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028472.5 | NP_001023643.1 | 8 - 9 | 1681 | 91 | |
NM_001028473.5 | NP_001023644.1 | 8 - 9 | 1677 | 91 | ||
NM_072013.8 | NP_504414.4 | 8 - 9 | 1681 | 91 | ||
NM_072014.7 | NP_504415.2 | 8 - 9 | 1690 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |