Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072018.8 | NP_504419.1 | 2 - 3 | 272 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |