Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072051.7 | NP_504452.1 | 2 - 3 | 870 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |