Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072054.6 | NP_504455.1 | 8 - 9 | 1387 | 92 | |
GenBank mRNA | AY204170.1 | - | 8 - 9 | 1254 | 92 | |
U41296.1 | - | 8 - 9 | 1285 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |