Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129372.4 | NP_001122844.1 | 1 - 2 | 189 | 71 | |
NM_072150.7 | NP_504551.3 | 1 - 2 | 235 | 71 | ||
GenBank mRNA | U09669.1 | - | 1 - 2 | 118 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |