Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028621.5 | NP_001023792.1 | 3 - 4 | 326 | 93 | |
NM_001380661.1 | NP_001367760.1 | 3 - 4 | 353 | 93 | ||
NM_001380662.1 | NP_001367759.1 | 3 - 4 | 405 | 93 | ||
NM_001383316.1 | 4 - 5 | 443 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |