Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028621.5 | NP_001023792.1 | 5 - 6 | 565 | 105 | |
NM_001380661.1 | NP_001367760.1 | 5 - 6 | 592 | 105 | ||
NM_001380662.1 | NP_001367759.1 | 5 - 6 | 644 | 105 | ||
NM_001383316.1 | 6 - 7 | 682 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |