Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpy-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383321.1 | 2 - 3 | 311 | 95 | ||
GenBank mRNA | AF250045.1 | - | 1 - 2 | 158 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |