Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dre-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380672.1 | NP_001368566.1 | 8 - 9 | 2691 | 79 | |
NM_001380673.1 | NP_001367735.1 | 8 - 9 | 2700 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |