Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072288.6 | NP_504689.1 | 12 - 13 | 1682 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |