Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028960.5 | NP_001024131.1 | 10 - 11 | 1329 | 80 | |
NM_072288.6 | NP_504689.1 | 9 - 10 | 1192 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |