Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356673.4 | NP_001343696.1 | 36 | 11799 | 83 | |
NM_001356674.3 | NP_001343811.1 | 36 | 11802 | 83 | ||
NM_001356675.3 | NP_001343689.1 | 37 | 11841 | 83 | ||
NM_001356676.3 | NP_001343809.1 | 37 | 11844 | 83 | ||
NM_001356677.4 | NP_001343695.1 | 38 | 12147 | 83 | ||
NM_001356678.4 | NP_001343810.1 | 38 | 12150 | 83 | ||
NM_001356679.3 | NP_001343688.1 | 39 - 40 | 12278 | 83 | ||
NM_001356680.4 | NP_001343808.1 | 39 | 12192 | 83 | ||
NM_001356681.3 | NP_001343815.1 | 36 - 37 | 11793 | 83 | ||
NM_001356682.3 | NP_001343819.1 | 36 - 37 | 11796 | 83 | ||
NM_001356683.3 | NP_001343813.1 | 37 - 38 | 11835 | 83 | ||
NM_001356684.3 | NP_001343817.1 | 37 - 38 | 11838 | 83 | ||
NM_001356685.3 | NP_001343814.1 | 38 - 39 | 12141 | 83 | ||
NM_001356686.3 | NP_001343818.1 | 38 - 39 | 12144 | 83 | ||
NM_001356687.3 | NP_001343812.1 | 39 - 40 | 12183 | 83 | ||
NM_001356688.3 | NP_001343816.1 | 39 - 40 | 12186 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |