Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322563.1 | NP_001309513.1 | 2 - 3 | 342 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |