Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072435.7 | NP_504836.1 | 7 - 8 | 3029 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |