Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072545.5 | NP_504946.2 | 3 - 4 | 385 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |