Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hint-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269160.3 | NP_001256089.1 | 3 - 4 | 291 | 62 | |
NM_001269161.2 | NP_001256090.1 | 2 - 3 | 202 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |