Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072697.5 | NP_505098.1 | 6 - 7 | 994 | 93 | |
GenBank mRNA | AF548624.1 | - | 6 - 7 | 994 | 93 | |
AY160228.1 | - | 6 - 7 | 991 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |